Anti-MCCC1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MCCC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-238
- 77833-237
- Antigen symbol:
- MCCC1
- MCCC1
- Antigen name:
- Methylcrotonoyl-CoA carboxylase subunit alpha, mitochondrial protein (1-200AA)
- Methylcrotonoyl-CoA carboxylase subunit alpha, mitochondrial protein (1-200AA)
- Antigen synonyms:
- MCCC1| MCCAMethylcrotonoyl-CoA carboxylase subunit alpha| mitochondrial| MCCase subunit alpha| EC 6.4.1.4| 3-methylcrotonyl-CoA carboxylase 1| 3-methylcrotonyl-CoA carboxylase biotin-containing subunit| 3-methylcrotonyl-CoA:carbon dioxide ligase subunit alpha
- MCCC1| MCCAMethylcrotonoyl-CoA carboxylase subunit alpha| mitochondrial| MCCase subunit alpha| EC 6.4.1.4| 3-methylcrotonyl-CoA carboxylase 1| 3-methylcrotonyl-CoA carboxylase biotin-containing subunit| 3-methylcrotonyl-CoA:carbon dioxide ligase subunit alpha
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96RQ3
- Q96RQ3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together