ESD Safe Solvent Dispenser, R & R Lotion
56614-114
Small Business Enterprise
- Capacity:177 ml (6 oz.)
- Description:ESD Safe Solvent Dispenser, with Purity Pump
- Cat. no.:56614-114
- Supplier No.:SD-6-ESD-AS
ESD safe solvent dispenser bottle with stainless steel anti-splash/purity pump.
- Blue ESD container with stainless steel pump
- Designed for use around static sensitive electronics
- Will not hold a charge
Cleanroom blue static dissipative solvent containers have an average surface resistivity of 10⁹ to 10¹⁰. All blue solvent dispensers will dissipate a charge of +/- 5000 volts in less than 2 seconds at 40 percent relative humidity. By their chemical nature, they will not have any chemical reactivity with solvents such as TCE or alcohol. Containers help to meet ANSI/ESDA S20.20.