ContactUsLinkComponent
Sterile IPA 70% Bulk, ITW Texwipe
Catalogue # CATWTX3290
Fournisseur:  ITW Texwipe
Sterile IPA 70% Bulk, ITW Texwipe
Catalogue # CATWTX3290
Fournisseur:  ITW Texwipe
Supplier Number:  TX3290
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Poly Bottle
  • Size:
    1 gal.
  • Cat. no.:
    CATWTX3290
  • Supplier No.:
    TX3290
Spécifications
À propos de cet article
This Sterile 70% Isopropanol (IPA) contains 70% by volume USP-grade Isopropanol and 30% USP-purified water
Wide down for pass through to controlled environments.
For facilities currently blending and using isopropanol solutions.
To ensure sterility, this solution is submicron filtered, filled into cleaned containers, double-bagged and gamma-irradiated. Each bottle is lot coded with an expiration date for easy recordkeeping.
Certifications: IPA quality meets requirements of USP Isopropyl Alcohol. Water quality meets requirements of USP Purified Water.