Flask, Round Bottom, Microscale, With Side Port, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Microscale, single-neck, round-bottom reaction flask
The top joint is a combination cap, thread and [ST] joint to accommodate and seal other microscale apparatus. Has an externally threaded side port to facilitate retrieval or injections. The O.D. of the top joint has an external thread to facilitate an o-ring and open-top phenolic cap. The internal or I.D. at the top is a ground outer joint. This type of connection eliminates the need for clamps while offering the positive leak-tight seal of ground glass. Fabricated from borosilicate glass.
Ordering information: Supplied with threaded cap with hole (89075-248 for top joint, 89075-244 for side port), PTFE-faced silicone rubber septum (89064-076 for top joint, 89064-072 for side port), and o-ring (89060-994 for top joint, 89060-596 for side port) for both top joint and side port.
Frequently Bought Together