Receiver, Moisture Test, Bidwell and Sterling-Type, Ace Glass
Catalog # 89060-336
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Capacity:5 ml
- Environmentally Preferable:
- Cat. no.:89060-336
- ST Joint Size:24/40
- Supplier No.:7705-02
Specifications
About this item
This Bidwell and Sterling style moisture test receiver is used to determine the moisture content in foods, fatty acids, etc
Fabricated from borosilicate glass.
Certifications: Conforms to specifications of ASTM and A.O.A.C.