Accessories for Brushes, Scratch, Replaceable Tips, Excelta Corp®
Supplier:   Excelta
Accessories for Brushes, Scratch, Replaceable Tips, Excelta Corp®
Supplier:  Excelta
Certificates
Accessories for Brushes, Scratch, Replaceable Tips, Excelta Corp®
Accessories for Brushes, Scratch, Replaceable Tips, Excelta Corp®
6 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Désignation
    Bristle Material
    Brush Head Width
    Availability
    Price
  • Replacement tip for 267, 2 star quality
    Fiberglass
    0.13" (3.25 mm)
    Log in for availability
    Log in for availability
    Each
    Each - $5.56
    $5.56
    Each
       $5.56
    Supplier Number: 267A
  • Replacement tip for 264, 2 star quality
    Fiberglass
    0.05" (1.25 mm)
    Log in for availability
    Log in for availability
    Each
    Each - $4.49
    $4.49
    Each
       $4.49
    Supplier Number: 264A
  • Replacement tip for 263, 2 star quality
    Fiberglass
    0.32" (8 mm)
    Log in for availability
    Log in for availability
    Each
    Each - $54.97
    $54.97
    Each
       $54.97
    Supplier Number: 263A
  • Replacement tip for 265, 2 star quality
    Brass
    0.13" (3.25 mm)
    Log in for availability
    Log in for availability
    Each
    Each - $4.74
    $4.74
    Each
       $4.74
    Supplier Number: 265A
  • Replacement tip for 266, 2 star quality
    Steel
    0.13" (3.25 mm)
    Log in for availability
    Log in for availability
    Each
    Each - $5.23
    $5.23
    Each
       $5.23
    Supplier Number: 266A
  • Replacement tip for 268, 2 star quality
    Nylon
    0.13" (3.25 mm)
    Log in for availability
    Log in for availability
    Each
    Each - $4.74
    $4.74
    Each
       $4.74
    Supplier Number: 268A