Wire Gowning Rack, Freestanding, With Hooks, Eagle MHC | Avantor
Wire Gowning Rack, Freestanding, With Hooks, Eagle MHC
Supplier:   Eagle Group
Wire Gowning Rack, Freestanding, With Hooks, Eagle MHC
Supplier:  Eagle Group
Certificates
Wire Gowning Rack, Freestanding, With Hooks, Eagle MHC
Wire Gowning Rack, Freestanding, With Hooks, Eagle MHC
3 Options Available Below
Product Details & Documents
Freestanding gowning rack incorporates wire shelving components that offer a more economical rack option for less demanding environments.
  • Convenient for storage of lab coats, uniforms and garments
  • Open wire construction facilitates laminar flow
Hooks on all four shelf sides are spaced on 1½" centers and are staggered 1" vertically hook-to-hook for ease of access.
Product Family Options
Informations sur le produit
  • Désignation
    Finish
    Matière
    Largeur
    Longueur
    Hauteur
    Availability
    Price
  • Free-standing gown rack
    Electropolished
    Stainless steel
    48"
    14"
    86"
    Discontinued
    Supplier Number: -
  • Free-standing gown rack
    Electropolished
    Stainless steel
    60"
    14"
    86"
    Discontinued
    Supplier Number: -
  • Free-standing gown rack
    Electropolished
    Stainless steel
    72"
    14"
    86"
    Discontinued
    Supplier Number: -
  • Free-standing gown rack
    Chrome
    Stainless steel
    48"
    14"
    86"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,260.23
    $4,260.23
    Each
       $4,260.23
    Supplier Number: C1448-GRH
  • Free-standing gown rack
    Chrome
    Stainless steel
    60"
    14"
    86"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,437.69
    $4,437.69
    Each
       $4,437.69
    La quantité minimale de commande est de 1
    Supplier Number: C1460-GRH
  • Free-standing gown rack
    Chrome
    Stainless steel
    72"
    14"
    86"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,325.19
    $5,325.19
    Each
       $5,325.19
    Supplier Number: C1472-GRH