VWR® Modular Workstations, W×D×H 48×30×36"
VWR® Modular Workstations, W×D×H 48×30×36"
VWR® Modular Workstations, W×D×H 48×30×36"
VWR® Modular Workstations, W×D×H 48×30×36"
VWR® modular work benches are designed for a wide range of applications in laboratory and industrial environments including biotech, pharmaceutical, analytical, material handling, distribution, and assembly.
  • Can be custom configured
  • Shipped fully assembled
  • Includes a variety of attachments
Standard work benches handle 1000lbs
The VWR® Basic Bench includes 4 standard leveler bench feet, gussets and light neutral powder coat of 16 gauge 1.5x1.5” steel frame. Shipped fully assembled. Also available in 60" and 72" widths.
of evenly distributed weight but can be configured to handle loads of 5000lbs. or more. Tables can be custom configured with numerous accessories including, 8" adjustable leg kits (for ADA compliance), locking casters, heavy-duty 2x2 steel frame, upper and lower shelves with pencil lips, backsplashes, lighting, bin rails, power strips, ESD tops, and steel drawers.
Ordering information: Select work bench size followed by type of work surface (epoxy, plastic laminate, ESD, phenolic). Next add appropriate accessories to the bench to complete your configuration. If you need something not on the price list, then please contact your Avantor sales representative. Exact frame dimensions depend upon reveal and thickness of work surface to be added.
Product Family Options
  • Désignation
    Hauteur
    W×D
  • Basic Bench
    36"
    48×30"
    Each
    Each - $1,987.25
    $1,987.25
    Each
       $1,987.25
  • Basic Bench
    36"
    48×30"
    Each
    Each - $4,508.51
    $4,508.51
    Each
       $4,508.51
  • Basic Bench
    36"
    48×30"
    Each
    Each - $5,002.98
    $5,002.98
    Each
       $5,002.98
  • Basic Bench
    36"
    48×30"
    Each
    Each - $3,559.61
    $3,559.61
    Each
       $3,559.61
  • Basic Bench
    36"
    48×30"
    Each
    Each - $2,821.41
    $2,821.41
    Each
       $2,821.41
Accessories
  •  
    Description
  • VF-ASD12
    Steel file drawer
    Each - $1,444.77
  •  
    Description
  • VF-ASD3
    Steel drawer
    Each - $990.42
  •  
    Description
  • VF-ASD6
    Steel drawer
    Each - $1,117.56
  •  
    Description
  • VF-A2FU
    Heavy Duty Steel Frame Upgrade, Depth 32", For Weight Loads over 1000 lbs.
    Each - $709.82
  •  
    Description
  • VF-AALK2
    Adjustable leg kit, premium, depth 36"
    Each - $1,278.11
  •  
    Description
  • VF-AALK1
    Adjustable leg kit, premium, depth 63"
    Each - $798.85
  •  
    Description
  • VF-ACTL
    Casters, Total Lock, Set of 4, Depth 36", Replaces Standard Leveler Bench Feet
    Each - $443.69
  •  
    Description
  • VF-ALSFL48
    Lower shelfdepth 28"
    Each - $683.09
  •  
    Description
  • VF-ALSFP48
    Lower shelf, depth 28"
    Each - $1,334.98
  •  
    Description
  • VF-AP48
    Powerstrip with 12 outlets, placed above or below the work surface
    Each - $871.66