Excelta Corp® Roto-Pic™ Vacuum Pickup System
Catalog # 63041-436
Supplier: Excelta
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Vacuum pickup system, complete kit, 5 star quality
- Cat. no.:63041-436
- Supplier No.:2000RK
Specifications
About this item
This system is Ideal for lifting, placing and turning components with the simple movement of one finger.
- Index finger slide switch for on/off
- Index finger wheel rotates tip 360°
- Available with many tip size/configurations
The 2000RK five star vacuum system is the most revolutionary vacuum pick-up available. Designed to reduce manual handling of both chips and wafers, the Roto-Pic has a smooth positive hold-and-release slide bar. The round nose wheel rotates 360° so components can be picked up and aligned with ease.
Ordering information: Includes Roto-Pic 2000A, 2000B, 2000C, 2020, 2021, 2022, 2023, 2024, 2050, 2051, 2052, 2060, 2071, 2040, 2041, 2043, 2044, 2080. Spare components are available at accessories.