Safety Enclosures by Labconco, METTLER TOLEDO®
Fournisseur: Mettler Toledo
Certificats
À propos de cet article
METTLER TOLEDO Enclosures by Labconco are specifically designed to protect the analyst during the weighing process
- Protection during weighing
- Clean-Sweep™ air foil
- Maximize containment
- Ensure safety
The patented Clean-Sweep™ air foil will maximize the containment of powders, particulates, and fumes to ensure operator safety. The ergonomic design and interior dimensions make this product ideal for micro and analytical balances, and the granite work surface will provide the greatest stability for your products.
Spécifications
- Catalog No:
- 89204-582
- 89174-754
- 89174-752
- Electrical:
- 115V, 60 Hz
- 115V, 60 Hz
- 115V, 60 Hz
Spécifications
Informations connexes
Frequently Bought Together