Ultimate FX Full Facepiece Air Purifying Respirator FF-400, 3M
89135-090
: 3M
- Size:Medium
- Cat. no.:89135-090
- Supplier No.:FF-402
Designed for flexibility in extreme environments.
- Head harness, comfort cradle, and soft silicone nose cup allow secure, comfortable wear over extended periods
- Large lens provides superior peripheral vision
- Bonded silicone gaskets and easy-to-use respirator cartridge/filter attachments
Scotchgard™ protector helps lens resist dirt, paint, and stains. Cool Flow™ exhalation valve reduces heat and moisture buildup inside facepiece.