Hand Sink, Pedestal Base, Pedal Valve, Eagle MHC
71001-946
- Bowl Dimensions (W×L×H):9¾×13½×6¾"
- Cat. no.:71001-946
- Supplier No.:HSA-10-FA-1P
Hand Sink is made up of Type 304 stainless steel.
- Heavy gauge
It features pedestal base with splash mounted faucet, single foot pedal valve, P-trap, tail piece and basket drain.