Sink, Mop, Floor Mount, Eagle MHC
Sink, Mop, Floor Mount, Eagle MHC
Supplier:   Eagle Group
Sink, Mop, Floor Mount, Eagle MHC
Supplier:  Eagle Group
Certificates
Sink, Mop, Floor Mount, Eagle MHC
Sink, Mop, Floor Mount, Eagle MHC
2 Options Available Below
Product Details & Documents
This sink has deep drawn one piece seamless construction of heavy gauge, type 304 stainless steel.
  • Anti-splash double-offset 'V' edge prevents spillage
  • Full skirt allows for tight sanitary seal
Water supply is ¹/₂" IPS for hot and cold lines. 2" drain.
Product Family Options
Product Information
  • Largeur
    Profondeur
    Longueur
    Availability
    Price
  • 16"
    8"
    20"
    Log in for availability
    Log in for availability
    Each
    Each - $3,953.07
    $3,953.07
    Each
       $3,953.07
    Minimum order quantity is 1
    Supplier Number: F1916-6
  • 28"
    12"
    20"
    Log in for availability
    Log in for availability
    Each
    Each - $5,488.63
    $5,488.63
    Each
       $5,488.63
    Minimum order quantity is 1
    Supplier Number: F2028-12