ContactUsLinkComponent
E.Z.N.A.® Tissue DNA Extraction Systems, Omega Bio-tek
undefined
E.Z.N.A.® Tissue DNA Extraction Systems, Omega Bio-tek
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Bacteria
    Blood
    Cultured cells
    FFPE tissues
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Sample size:
    30 mg or 5 x 10⁶ Cells
  • Description:
    Tissue DNA Kit
Frequently Bought Together