Drop In Sink, Self Rim Design, Three Bowl, Eagle MHC
Heavy gauge type 304 series stainless steel construction.
- Sinks are deep-drawn and self rimming
- Positive holddown clamping tabs for top mount
Faucet holes punched on 4" center.
: Faucet and drain, mounting hardware included.
Frequently Bought Together