VK Sponge, Contec® | Avantor
VK Sponge, Contec®
VK Sponge, Contec®
Supplier:  CONTEC, INC. MX
Certificates
VK Sponge, Contec®
VK Sponge, Contec®
2 Options Available Below
Product Details & Documents
The VertiKlean® Sponge is a disposable cleanroom sponge that features high sorbency polyester foam laminated to a Polynit wiping material with excellent abrasion resistance.
  • High sorbency polyester foam makes sponges very sorbent
  • Textured polyester surface for more aggressive cleaning tasks
  • The sponge is laser cut to completely seal the edges
Designed for general cleaning and wiping, they are ideal for use in the laboratory. Autoclavable or supplied as a sterile, ready-to-use product.
Packaging: Non sterile sponges are packaged 8/bag, 3 bags/inner bag, 4 inner bags/case. Sterile sponges are packaged 1/bag, 15 bags/inner bag, 8 inner bags/case.
Product Family Options
Informations sur le produit
  • Désignation
    L×W×H
    Availability
    Price
  • VK Sponge, Single-Ply, Sterile
    12.7×12.7×1.5 cm (5×5×0.6")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 120
    Case of 120 - $1,565.20
    $1,565.20
    Prix promotionnel pour les nouveaux clients
    Case of 120
       $1,565.20
    Supplier Number: VKST0003
  • VK Sponge, Single-Ply, Non sterile
    22×11×1.5 cm (8.6×4.3×0.6")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 8
    Pack of 8 - $63.04
    $63.04
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 96
    Case of 96 - $756.46
    $756.46
    Prix promotionnel pour les nouveaux clients
    Pack of 8
       $63.04
    Case of 96
       $756.46
    Supplier Number: VKSPONGE-1