WHEATON® MicroLiter µLPlate® Component Kits, PP Plates with Covers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
MicroLiter µLPlate® components include a proprietary product line of 96-well round and square well microplates with covers.
- Variety of volumes available
- EVA covers offer superior seal
- Silicone mats are resealable
The 96-well round and square well microplates are available in a variety of volumes. Constructed of natural polypropylene, all microplates are made to ANSI specifications for 96-well microplates. Covers made of Ethylene Vinyl Acetate (EVA) are piercible by standard needle configurations found on most HPLC, LC/MS, CombiChem, and HTS instruments. They have a superior seal compared to similar products. For users who require some level of resealability, two silicone mats with a sprayed-on fluoropolymer resin barrier are available. Silicone mats are available in standard and pre-cut formats.
Frequently Bought Together