AdjusTable® Workstation, Stainless Steel Perforated Top, Stainless Steel Base, Eagle MHC™
Type 304 perforated brushed stainless steel top.
- 14 gauge worksurface with ¾" perforations on 1" centers
- Stainless steel base feature four 33" stainless posts
- One stainless wire shelf and stainless C-frame
Load capacity: 600 lbs.
Frequently Bought Together