AdjusTable® Workstation, Stainless Steel Solid Top, Chrome Base, Eagle MHC™
AdjusTable® Workstation, Stainless Steel Solid Top, Chrome Base, Eagle MHC™
AdjusTable® Workstation, Stainless Steel Solid Top, Chrome Base, Eagle MHC™
AdjusTable® Workstation, Stainless Steel Solid Top, Chrome Base, Eagle MHC™
16 gauge, Type 304 brushed stainless steel top. All work surface tabletops feature hat channel underneath for added strength.
  • 34.5" (876 mm) working height
  • Easy to add undershelves or overshelves
  • Available in solid or perforated, with brushed or electropolished finish
  • Four 33" (838 mm) chrome posts or stainless steel posts numbered in 1" (25 mm) increments for flexible and level positioning of the base
Base combination available with or without wire undershelf, with chrome or stainless steel H-frame(s) or C-frame(s).
Load capacity: 600 lbs.
Product Family Options
  • Désignation
    Capacité
    Worksurface Material
    Base/Leg Material
    Hauteur
    W×D
    Shipping Weight
  • Workstation with solid top
    600 lbs.
    Stainless steel
    Stainless steel
    34½"
    36×24"
    65 lbs.
    Each
    Each - $2,614.67
    $2,614.67
    Each
       $2,614.67
     : AC2436T
  • Workstation with solid top
    600 lbs.
    Stainless steel
    Stainless steel
    34½"
    48×24"
    81 lbs.
    Each
    Each - $3,121.81
    $3,121.81
    Each
       $3,121.81
     : AC2448T
  • Workstation with solid top
    600 lbs.
    Stainless steel
    Stainless steel
    34½"
    60×24"
    97 lbs.
    Each
    Each - $3,521.92
    $3,521.92
    Each
       $3,521.92
     : AC2460T
  • Workstation with solid top
    600 lbs.
    Stainless steel
    Stainless steel
    34½"
    36×30"
    75 lbs.
    Each
    Each - $2,794.15
    $2,794.15
    Each
       $2,794.15
     : AC3036T
  • Workstation with solid top
    600 lbs.
    Stainless steel
    Stainless steel
    34½"
    48×30"
    94 lbs.
    Each
    Each - $3,451.54
    $3,451.54
    Each
       $3,451.54
     : AC3048T
  • Workstation with solid top
    600 lbs.
    Stainless steel
    Stainless steel
    34½"
    60×30"
    113 lbs.
    Each
    Each - $3,907.96
    $3,907.96
    Each
       $3,907.96
     : AC3060T