Stainless Steel Worktable with Backsplash and Tubular Base, 16-Gauge Type 304, Eagle MHC™
Supplier: Eagle Group
Certificates
About this item
16 gauge type 304 stainless steel top with 4" backsplash at rear of table.
- 35¹/₈" to 36¹/₈" adjustable working height
- 1⁵/₈" diameter legs and 1¹/₄" diameter crossbracing are heavy gauge stainless steel
- Rolled edge front with ends turned under 90 degrees
.
Ordering information: Accessories available include drawers and casters, sold separately.
Frequently Bought Together