Wipe Dispenser with Bottle Holder, Stainless Steel, Bandy
89217-404
: Bandy
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- Description:Bottle Dispenser
- Dimensions:16.4"Wx4.3"Dx12.3"H
- Cat. no.:89217-404
- Supplier No.:DISP11007
Wiper dispenser with bottle holder. Made from stainless steel 304.
- Electropolished
- Perforated Bottom
- Ships Assembled
Holds 4" Diameter Bottle. Wipe Compartment 4"x12".