ContactUsLinkComponent
STERILE VERTEX® Polyester Pre-Wetted Wipers, Texwipe®
STERILE VERTEX® Polyester Pre-Wetted Wipers, Texwipe®
Supplier:  Texwipe
Certificates
undefined
STERILE VERTEX® Polyester Pre-Wetted Wipers, Texwipe®
Supplier:  Texwipe
Certificates
Specifications
  • Catalog No:
  • TWTX3049P
  • TWTX3042P
  • Absorbency:
  • 450 ml/m²
  • 450 ml/m²
  • Fibres (>100 µm):
  • 200/m²
  • 200/m²
  • Particle generation (>=0,5 µm):
  • 9.5 x 10⁶/m²
  • 9.5 x 10⁶/m²
  • Particle generation (0,5 - 5 µm) - ASTM E2090 test:
  • 3.0 x 10⁶/m²
  • 3.0 x 10⁶/m²
  • Particle generation (5 - 100 µm) - ASTM E2090 Test:
  • 66,000/m²
  • 66,000/m²
  • Ions - calcium:
  • <0.10 ppm
  • <0.10 ppm
  • Ions - chloride:
  • <0.05 ppm
  • <0.05 ppm
  • Ions - magnesium:
  • <0.01 ppm
  • <0.01 ppm
  • Ions - sodium:
  • <0.17 ppm
  • <0.17 ppm