NeuLog Soil Moisture Logger Sensor | Avantor
NeuLog Soil Moisture Logger Sensor
NeuLog soil moisture logger sensor, 0 - 50 Kpa, 0.02 Kpa
ADC: 10 bit
Catalog # 470137-424
Supplier:   Wards
Fournisseur #:  NUL229
NeuLog Soil Moisture Logger Sensor
Catalog # 470137-424
Supplier:  Wards
NeuLog Soil Moisture Logger Sensor
NeuLog Soil Moisture Logger Sensor
NeuLog soil moisture logger sensor, 0 - 50 Kpa, 0.02 Kpa
ADC: 10 bit
Price
$276.45
Log in pour la disponibilité
Each
Product Details & Documents
A unique approach to measuring moisture via vacuum pressure in a tensiometer.
  • Max. sample rate 100 S/sec.
  • Experimental duration 1 sec. to 31 days
The tensiometer on this logger sensor measures soil moisture via pressure level. When the soil is dry the water exits ceramic holes in the end due to diffusion, when the soil is wet the vacuum in the tensiometer pulls water inside and the vacuum decreases.
Ordering information: For each probe you will need one or more of the following items: Battery Module, Viewer module (not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
Caution: For educational use only.
Spécifications
  • Résolution:
    0.02 Kpa
    ADC: 10 bit
  • Désignation:
    NeuLog soil moisture logger sensor
  • Range:
    0 - 50 Kpa
  • Cat. No.:
    470137-424
  • Interface:
    USB
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • NeuLog USB module
    NeuLog USB module
    470137-442
    Log in pour la disponibilité
    Each - $119.20
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Neulog WiFi 2.0
    Neulog WiFi 2.0
    470221-194
    Log in pour la disponibilité
    Each - $268.90
Informations sur le produit
  • Catalog #
    Availability
    Price
  • NeuLog viewer graphic color display module
    NeuLog viewer graphic color display module
    470137-438
    Log in pour la disponibilité
    Each - $379.20