NeuLog Turbidity Logger Sensor | Avantor
NeuLog Turbidity Logger Sensor
NeuLog turbidity logger sensor, 0 - 200 NTU, 0.08 NTU
ADC: 10 bit
Catalog # 470137-428
Supplier:   Wards
Fournisseur #:  NUL231
NeuLog Turbidity Logger Sensor
Catalog # 470137-428
Supplier:  Wards
NeuLog Turbidity Logger Sensor
NeuLog Turbidity Logger Sensor
NeuLog turbidity logger sensor, 0 - 200 NTU, 0.08 NTU
ADC: 10 bit
Price
$338.75
Log in pour la disponibilité
Each
Product Details & Documents
Easily determine turbidity.
  • Max. sample rate 100 S/sec.
  • Experimental duration 1 sec. to 31 days
This sensor measures reflected light in order to determine solution turbidity. As the turbidity grows higher more light is reflected and measured in Nephelometric Turbidity Unit (NTU) by the light sensor which is located perpendicular to the tube.
 : For each probe you will need one or more of the following items: Battery Module, Viewer module(Not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
 : For educational use only.
Spécifications
  • Résolution:
    0.08 NTU
    ADC: 10 bit
  • Désignation:
    NeuLog turbidity logger sensor
  • Range:
    0 - 200 NTU
  • Cat. No.:
    470137-428
  • Interface:
    USB
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • NeuLog viewer graphic color display module
    NeuLog viewer graphic color display module
    470137-438
    Log in pour la disponibilité
    Each - $379.20
Informations sur le produit
  • Catalog #
    Availability
    Price
  • NeuLog USB module
    NeuLog USB module
    470137-442
    Log in pour la disponibilité
    Each - $119.20
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Neulog WiFi 2.0
    Neulog WiFi 2.0
    470221-194
    Log in pour la disponibilité
    Each - $268.90