Axygen® AxyPrep™ Mag Dye Clean-Up Kit, Corning
Supplier:   Corning
Axygen® AxyPrep™ Mag Dye Clean-Up Kit, Corning
Supplier:  Corning
Certificates
4 Options Available Below
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
AxyPrep™ Mag DyeClean utilizes a unique paramagnetic bead-based sequencing purification system optimized for the clean-up of excess dye terminator post sequencing reaction
  • Significant BigDye savings without incurring extra cost and processing time
  • High signal to noise ratio resulting in long reads and high QV20+ scores
  • Suitable for the clean-up of sequencing reaction of different Sanger sequencing based platforms
  • Simple and easy sequencing reaction clean-up process: 25 minutes/96 samples
  • No centrifugation or filtration required
Excess dye terminator clean-up post sequencing reaction clean-up is an essential step prior to sequencing. The carry over of excess dye into sequencing results in dye blobs which leads to overwhelming noise and failed calls of sequenced bases, potentially leading to the failure of the sequencing run. AxyPrep™ Mag DyeClean utilizes a unique paramagnetic bead-based sequencing purification system optimized for the clean-up of excess dye terminator post sequencing reaction in Sanger sequencing. The protocol mainly consists of binding, washing, and elution steps, and can be performed directly in the thermal cycling plate, requiring no centrifugation or filtration.
The protocol mainly consists of binding, washing, and elution steps, and can be performed directly in the thermal cycling plate. It requires no centrifugation or filtration.
Each insert has its own electrode assembly that connects to the lid of the buffer tank. Coiled electrodes within the insert generate a high intensity current, allowing standard transfer to take place in approximately one hour. Power leads connect directly to lid so that power is disconnected from system when the lid is removed. Buffer tank is molded to prevent separation and leaking.
 : System includes buffer tank with power leads and cooling pack; blotting insert; three blotting cassettes; six fiber pads; two 10x10 cm (4x4") notched plates; two 10x10 cm (4x4") plates with spacers; and two 12-tooth, 1mm (1/32") thick combs.
Specifications for Product Family
Spécifications
Spécifications
  • Catalog No:
  • 10159-710
  • 97067-459
  • 97067-461
  • 97067-471
  • Description:
  • AxyPrep™ Mag DyeClean Up Kit
  • AxyPrep™ Mag DyeClean Up Kit
  • AxyPrep™ Mag DyeClean Up Kit
  • AxyPrep™ Mag DyeClean Up Kit
  • Target molecule:
  • DNA
  • DNA
  • DNA
  • DNA
  • Refine target molecule:
  • Amplified/modified DNA
  • Amplified/modified DNA
  • Amplified/modified DNA
  • Amplified/modified DNA
  • Sample type:
  • Sequencing reactions/libraries
  • Sequencing reactions/libraries
  • Sequencing reactions/libraries
  • Sequencing reactions/libraries
  • Method / Format:
  • Magnetic beads
  • Magnetic beads
  • Magnetic beads
  • Magnetic beads
  • Prep size:
  • Scalable
  • Scalable
  • Scalable
  • Scalable
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Volume
    Taille
    Availability
    Price
  • AxyPrep™ Mag DyeClean Up Kit
    1 ml
    200 Preps
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $224.00
    $224.00
    Log in pour la disponibilité
    Each
    Each - $224.00
    $224.00
    Case of 1
       $224.00
    Each
       $224.00
    Supplier Number: MAG-DYECL-1
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    10159-710
    Description
    AxyPrep™ Mag DyeClean Up Kit
    Target molecule
    DNA
    Refine target molecule
    Amplified/modified DNA
    Sample type
    Sequencing reactions/libraries
    Method / Format
    Magnetic beads
    Prep size
    Scalable
  • AxyPrep™ Mag DyeClean Up Kit
    5 ml
    500 Preps
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $470.00
    $470.00
    Log in pour la disponibilité
    Each
    Each - $470.00
    $470.00
    Case of 1
       $470.00
    Each
       $470.00
    Supplier Number: MAG-DYECL-5
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    97067-459
    Description
    AxyPrep™ Mag DyeClean Up Kit
    Target molecule
    DNA
    Refine target molecule
    Amplified/modified DNA
    Sample type
    Sequencing reactions/libraries
    Method / Format
    Magnetic beads
    Prep size
    Scalable
  • AxyPrep™ Mag DyeClean Up Kit
    50 ml
    5000 Preps
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $2,590.00
    $2,590.00
    Log in pour la disponibilité
    Each
    Each - $2,590.00
    $2,590.00
    Case of 1
       $2,590.00
    Each
       $2,590.00
    Supplier Number: MAG-DYECL-50
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    97067-461
    Description
    AxyPrep™ Mag DyeClean Up Kit
    Target molecule
    DNA
    Refine target molecule
    Amplified/modified DNA
    Sample type
    Sequencing reactions/libraries
    Method / Format
    Magnetic beads
    Prep size
    Scalable
    Product available on GSA Advantage®
  • AxyPrep™ Mag DyeClean Up Kit
    250 ml
    25000 Preps
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $28,510.00
    $28,510.00
    Case of 1
       $28,510.00
    Supplier Number: MAGDYECL-250
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    97067-471
    Description
    AxyPrep™ Mag DyeClean Up Kit
    Target molecule
    DNA
    Refine target molecule
    Amplified/modified DNA
    Sample type
    Sequencing reactions/libraries
    Method / Format
    Magnetic beads
    Prep size
    Scalable