Bottles, Square, High-Density Polyethylene, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
Leakproof, chemical-resistant bottles have heavy threaded necks and tamper-proof, tear-lock closures.
- Chemical-resistant
- Heavy-threaded necks
- HDPE construction
- Leakproof
Molded of high-density polyethylene with deep-shelled, blue polypropylene closures, they are ideal for storing or shipping samples.
Frequently Bought Together