ClipTip® Pipette Tips, 200
ClipTip® pipette tips provide security with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force.
All ClipTip® pipette tips feature a hydrophobic inner surface which effectively prevent liquid retention. ClipTip® tips are certified free of RNase, DNase, DNA, ATP, and pyrogen contamination. The tips are available in five volumes and in multiple packaging options, including environmentally friendly reload stacks.
ClipTip® 200 pipette tips suit pipetting of 10 to 200 µl volumes.
The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip® pipette until – and only until – it is released.
: Manufactured according to ISO 9001 and ISO 13485. The F1-ClipTip® pipetting system is CE-marked in accordance with IVD Directive in Europe.
ClipTip® pipette tips provide security with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force.
All ClipTip® pipette tips feature a hydrophobic inner surface which effectively prevent liquid retention. ClipTip® tips are certified free of RNase, DNase, DNA, ATP, and pyrogen contamination. The tips are available in five volumes and in multiple packaging options, including environmentally friendly reload stacks.
ClipTip® 200 pipette tips suit pipetting of 10 to 200 µl volumes.
The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip® pipette until – and only until – it is released.
: Manufactured according to ISO 9001 and ISO 13485. The F1-ClipTip® pipetting system is CE-marked in accordance with IVD Directive in Europe.
Product Family Options
- Tip StyleVolumeSterileFilterLongueurTip ColorFor Use WithPacked
- ClipTip® 200, Racked10 - 200 µl5.6 cmJauneF1-ClipTip® 10 - 100 μl, 20 - 200 μl, Fixed 100 and 200 μl, 8 - Channel 10 - 100 μl, and 12 - Channel 10 - 100 μl10×96/RackCase of 960$109.37: 94410310New ProductNew Product
- ClipTip® 200, Racked10 - 200 µl+5.6 cmJauneF1-ClipTip® 10 - 100 μl, 20 - 200 μl, Fixed 100 and 200 μl, 8 - Channel 10 - 100 μl, and 12 - Channel 10 - 100 μl10×96/RackCase of 960$140.53: 94410313New ProductNew Product
- ClipTip® 200, Reload Stacks10 - 200 µl5.6 cmJauneF1-ClipTip® 10 - 100 μl, 20 - 200 μl, Fixed 100 and 200 μl, 8 - Channel 10 - 100 μl, and 12 - Channel 10 - 100 μl10×96/RackCase of 960$101.29: 94410317New ProductNew Product
- ClipTip® 200, Reload Stacks10 - 200 µl+5.6 cmJauneF1-ClipTip® 10 - 100 μl, 20 - 200 μl, Fixed 100 and 200 μl, 8 - Channel 10 - 100 μl, and 12 - Channel 10 - 100 μl10×96/InsertCase of 960$129.68: 94410318New ProductNew Product
- ClipTip® 200, Filter, Reload10 - 200 µl++5.6 cmJauneF1-ClipTip® 10 - 100 μl, 20 - 200 μl, Fixed 100 and 200 μl, 8 - Channel 10 - 100 μl, and 12 - Channel 10 - 100 μl10×96/InsertCase of 960$182.28: 94420318New ProductNew Product
- ClipTip® 200, Racked10 - 200 µl+5.6 cmJauneF1-ClipTip® 10 - 100 μl, 20 - 200 μl, Fixed 100 and 200 μl, 8 - Channel 10 - 100 μl, and 12 - Channel 10 - 100 μl10×96/Rack-: 94410313
- ClipTip® 200, Reload Stacks10 - 200 µl5.6 cmJauneF1-ClipTip® 10 - 100 μl, 20 - 200 μl, Fixed 100 and 200 μl, 8 - Channel 10 - 100 μl, and 12 - Channel 10 - 100 μl10×96/Insert-: -
- ClipTip® 200, Racked10 - 200 µl5.6 cmJauneF1-ClipTip® 10 - 100 μl, 20 - 200 μl, Fixed 100 and 200 μl, 8 - Channel 10 - 100 μl, and 12 - Channel 10 - 100 μl10×96/Rack-: 94410310
- ClipTip® 200, Racked10 - 200 µl+5.6 cmJauneF1-ClipTip® 10 - 100 μl, 20 - 200 μl, Fixed 100 and 200 μl, 8 - Channel 10 - 100 μl, and 12 - Channel 10 - 100 μl10×96/Rack-: 94410313
- ClipTip® 200, Reload Stacks10 - 200 µl+5.6 cmJauneF1-ClipTip® 10 - 100 μl, 20 - 200 μl, Fixed 100 and 200 μl, 8 - Channel 10 - 100 μl, and 12 - Channel 10 - 100 μl10×96/Insert-: 94410318
- ClipTip® 200, Filter, Reload10 - 200 µl++5.6 cmJauneF1-ClipTip® 10 - 100 μl, 20 - 200 μl, Fixed 100 and 200 μl, 8 - Channel 10 - 100 μl, and 12 - Channel 10 - 100 μl10×96/Insert-: 94420318
- ClipTip® 200, Reload Stacks10 - 200 µl5.6 cmJauneF1-ClipTip® 10 - 100 μl, 20 - 200 μl, Fixed 100 and 200 μl, 8 - Channel 10 - 100 μl, and 12 - Channel 10 - 100 μl10×96/Insert-: 94410317