Safety Glass Dispenser, 7 Compartments, S-Curve Technologies
Supplier: S-Curve Technologies
Certificates
About this item
Safety Glass Dispenser for 7 pairs of glasses.
- Mounts to wall vertically or horizontally
- Flame treated edges for smoothness
Made from clear acrylic or PETG.
Specifications
- Catalog No:
- 10703-482
- 89208-714
- PPE type:
- Safety glasses
- Safety glasses
- Material:
- PETG
- Acrylic
- No. of compartments:
- 7
- 7
- Dispenser type:
- Wall-mountable
- Wall-mountable
- Thickness:
- 6 mm (0.25")