NxGen™ M-MulV Reverse Transcriptase, Biosearch Technologies
NxGen™ M-MulV Reverse Transcriptase, Biosearch Technologies
Supplier:   Lucigen
NxGen™ M-MulV Reverse Transcriptase, Biosearch Technologies
Supplier:  Lucigen
Certificates
NxGen™ M-MulV Reverse Transcriptase, Biosearch Technologies
NxGen™ M-MulV Reverse Transcriptase, Biosearch Technologies
1 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Lucigen’s NxGen™ M-MulV Reverse Transcriptase is outstanding in cDNA synthesis.
M-MuLV Reverse Transcriptase is an RNA-dependent DNA polymerase which shows no measurable 3′→5′ proofreading function. This enzyme can copy a single-stranded DNA template or perform cDNA synthesis by extending a DNA primer annealed to an RNA template.
Performance Specifications: 1) Unit Concentration: 200,000 U/mL. 2) Purity (SDS-PAGE): >99%. 3) SS Exonuclease: 2,000 U <5.0% released. 4) DS Exonuclease: 2,000 U <0.5% released. 5) Endonuclease: 2,000 U <10% converted. 5) E. coli 16S rDNA Contamination: 2,000 U <10 copies. 6) Storage: -20°C.
Source: A recombinant E. coli strain carrying the Moloney-Murine Leukemia Virus Reverse Transcriptase gene.
Ordering information: Supplied at 200,000 U/mL in 50 mM Tris-HCl, 150 mM NaCl, 0.1 mM EDTA, 1 mM dithiothreitol, 0.1% NP-40 Alternative, 50% glycerol, pH 7.6 @ 25°C. Also provided is 10X M-MuLV RT Buffer which in 1X form is composed of 50 mM Tris-HCl, 75 mM KCl, 3 mM MgCl2, 10 mM dithiothreitol, pH 8.3 @ 25°C. 250,000 U is supplied as 5 × 50,000 U. Unit definition: 1 unit is defined as the amount of enzyme required to incorporate 1 nmol of dTTP into acid insoluble material in 10 minutes at 37°C using poly r(A)/oligo (dT) as a substrate.
Product Family Options
Product Information
  • Désignation
    Units
    No. of Reactions
    Availability
    Price
  • NxGen™ M-MulV Reverse Transcriptase
    50000
    50000
    Log in for availability
    Log in for availability
    Each
    Each - $758.01
    $758.01
    Each
       $758.01
    Minimum order quantity is 1
    Supplier Number: 30222-1
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • NxGen™ M-MulV Reverse Transcriptase
    250000
    250000
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Accessories
Product Information
  • Catalog #
    Size
    Availability
    Price
  • CA97065-224
    10,000 U
    Log in for availability
    Each - $1,857.00
Product Information
  • Catalog #
    Size
    Availability
    Price
  • CA97065-226
    50,000 U
    Log in for availability
    Each - $8,446.32