Total Suspended Solids (TSS) Standard, 100 mg/L (100 ppm) TSS (Gravimetric, Dried at 103 - 105 °C), Ricca Chemical Company
Catalog # RC8672-16
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Size:16 oz.
- Cat. no.:RC8672-16
- Supplier No.:R8672000-500C