Phosphate Buffer, for Cyanide, Ricca Chemical Company
Catalog # RC5810-32
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Size:32 oz.
- Cat. no.:RC5810-32
- Supplier No.:R5810000-1A
Specifications
About this item
Suitable for use in the following compendial methods: -ASTM D2036 D as Reagent 36.6 -APHA 4500-CN H as Reagent 4g