Electrode Cleaning Solution, for Removal of Membrane/Junction Protein Deposits, Ricca Chemical Company | Avantor
Electrode Cleaning Solution, for Removal of Membrane/Junction Protein Deposits, Ricca Chemical Company
Poly Bottle, 500 ml
Catalog # RC279416
Supplier:   Ricca Chemical
Fournisseur #:  R2794000-500A
Electrode Cleaning Solution, for Removal of Membrane/Junction Protein Deposits, Ricca Chemical Company
Catalog # RC279416
Supplier:  Ricca Chemical
CAS Number:  
Electrode Cleaning Solution, for Removal of Membrane/Junction Protein Deposits, Ricca Chemical Company
Electrode Cleaning Solution, for Removal of Membrane/Junction Protein Deposits, Ricca Chemical Company
Poly Bottle, 500 ml
CAS Number:  
Price
$203.43
Log in pour la disponibilité
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Electrode Cleaning Solutions for the Removal of Membrane/Junction Protein Deposits.
Shelf life: 12 months
Spécifications
Spécifications
  • Désignation:
    Poly Bottle
  • Taille:
    500 ml