Buffer Solution, for Nitrate Determination using Ion Selective Electrode, Ricca Chemical Company | Avantor
Buffer Solution, for Nitrate Determination using Ion Selective Electrode, Ricca Chemical Company
500 mL
Catalog # RC147316
Supplier:   Ricca Chemical
Fournisseur #:  R1473000-500C
Buffer Solution, for Nitrate Determination using Ion Selective Electrode, Ricca Chemical Company
Catalog # RC147316
Supplier:  Ricca Chemical
CAS Number:  
Buffer Solution, for Nitrate Determination using Ion Selective Electrode, Ricca Chemical Company
Buffer Solution, for Nitrate Determination using Ion Selective Electrode, Ricca Chemical Company
500 mL
CAS Number:  
Price
243,20 $
Log in pour la disponibilité
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Reagent 3d (Buffer Solutions) for APHA method 4500-NO3-, D: Nitrate Electrode Method
Spécifications
Spécifications
  • Taille:
    500 mL