Sealed Sample Tubes, Omni International
Sealed sample tubes are used with clear Omni Tip™ plastic generator probes to reduce harmful airborne contaminants.
- Sealed tubes are designed as a secondary means of protection
Always use appropriate measures when processing pathogens.
: Omni Tip™ Plastic Generator Probes are sold separately.
Frequently Bought Together