Double-wall weatherproof cabinets are all-welded, 18-gauge construction using galvannealed steel and painted with a weather-resistant, impact-resistant urethane finish that is also resistant to ultra-violet rays.
All Cabinets STANDARD Features a sloped roof with a 2³/₄" overhang along with interior and exterior astragals to prevent rain penetration. Weatherproof cabinets come standard with corrosion resistant flush-mounted stainless steel locking handle, but may be upgraded to galvanized high security hasp lock. Easily move these cabinets to the appropriate work location using built-in heavy gauge fork lift channels and secure them with fully-welded 7-gauge steel Bolt-Down Brackets with ⁷/₈" diameter opening. Available on all standard cabinet models with manual and self-closing door styles.
- Catalog No:
- 89237-756
- 89237-758
- 89237-744
- 89237-746
- 89237-748
- 89237-750
- 89237-752
- 89237-754
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified