Sterile TexTra™ 10 Polyester Wipers, Texwipe®
Sterile TexTra™ 10 Polyester Wipers, Texwipe®
Supplier:   Texwipe
Sterile TexTra™ 10 Polyester Wipers, Texwipe®
Supplier:  Texwipe
Certificates
Sterile TexTra™ 10 Polyester Wipers, Texwipe®
Sterile TexTra™ 10 Polyester Wipers, Texwipe®
2 Options Available Below
Product Details & Documents
Sterile polyester wipers are effective for cleaning equipment and environmental surfaces in critical aseptic cleanrooms without compromising the sterile environment.
  • Single-ply
  • double-knit
  • Cleanroom laundered
Wipers are made of 100% continuous-filament polyester, have sealed borders.
Suitable for the final rinsing and residue removal step of most critical cleaning protocols.
Certifications: Gamma irradiated to a Sterility Assurance Level of 10⁻⁶. For use in ISO Class 3-5 cleanroom environments.
Ordering information: Double-bagged in solvent safe bags that can be wiped down with alcohol before introduction of inner bag to sterile environments.
Packaging: 500 wipers total/case.
Specifications for Product Family
Specifications
  • Catalog No:
  • TWTX3224
  • CATWTX3225
  • Absorbency:
  • 600 ml/m²
  • 600 ml/m²
  • Fibres (>100 µm):
  • 240/m²
  • 240/m²
  • Particle generation (0,5 - 5 µm) - ASTM E2090 test:
  • 8 x 10⁶/m²
  • 8 x 10⁶/m²
  • Particle generation (5 - 100 µm) - ASTM E2090 Test:
  • 220,000/m²
  • 220,000/m²
Specifications
Product Family Options
Product Information
  • W×L
    Packaging
    Availability
    Price
  • 22.9 x 22.9 cm (9 x 9")
    100 wipers/bag (5 inner bags of 20 wipers), 5 bags/case
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $888.00
    $888.00
    New Customer Promotional Price
    Case of 500
       $888.00
    Supplier Number: TX3224
    Specifications:
    More
    Specifications
    Cat. No.
    TWTX3224
    Absorbency
    600 ml/m²
    Fibres (>100 µm)
    240/m²
    Particle generation (0,5 - 5 µm) - ASTM E2090 test
    8 x 10⁶/m²
    Particle generation (5 - 100 µm) - ASTM E2090 Test
    220,000/m²
  • 31 x 31 cm (12 x 12")
    100 wipers/bag (5 inner bags of 20 wipers), 5 bags/case
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $3,080.00
    $3,080.00
    Case of 500
       $3,080.00
    Supplier Number: TX3225
    Specifications:
    More
    Specifications
    Cat. No.
    CATWTX3225
    Absorbency
    600 ml/m²
    Fibres (>100 µm)
    240/m²
    Particle generation (0,5 - 5 µm) - ASTM E2090 test
    8 x 10⁶/m²
    Particle generation (5 - 100 µm) - ASTM E2090 Test
    220,000/m²