Fused-Insert Vials, Screw Caps w/ Pre-Slit Septa, MicroSolv™
These vials have a small volume insert installed and permanently 'fused' in.
These vials/inserts are available for snap/crimp top, 9 mm screw top vials. The ultimate in convenience when using small volumes. These vials have screw caps with PTFE/Silicone septa.
Frequently Bought Together