Clearview Classroom Stream Table | Avantor
Clearview Classroom Stream Table
New Product
Supplier:   Science First
Clearview Classroom Stream Table
Supplier:  Science First
New Product
Certificates
Clearview Classroom Stream Table
Clearview Classroom Stream Table
1 Options Available Below
Product Details & Documents
Simulate a flowing stream and study erosion with this classroom stream table.
  • Clear surface for visual study
    Ideal for environmental and water studies
    Water tight
    Easy to store
Bring Water Dynamics to Life with the Clearview Classroom Stream Table - a must have for any earth science classroom! This interactive stream table offers a dynamic way for students to explore erosion, sediment transport, and river formation. Designed for easy setup and durability, the rugged ABS construction features an all-in-one system—just add water, activate the pump, and start experimenting. Five adjustable water jets simulate natural water flow, from small streams to converging trib. Ships oversize.
Product Family Options
Informations sur le produit
  • Désignation
    Width
    Longueur
    Availability
    Price
  • Clearview Classroom Stream Table
    20"
    48"
    Discontinued
    Supplier Number: 651-6215
  • Replacement pump
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $63.12
    $63.12
    Each
       $63.12
    Supplier Number: 035590
    Spécifications:
    New Product
    New Product