ContactUsLinkComponent
National 200 Series Compound Microscope
Catalog # 470136-760
Supplier:  National Optical
undefined
National 200 Series Compound Microscope
Catalog # 470136-760
Supplier:  National Optical
Supplier Number:  215-RLED
Specifications
  • Description:
    National 215 Compound Binocular Microscope, 4X, 10X, 40X, 100X, Mech. Stage
  • Cat. no.:
    470136-760
  • Illumination:
    LED Cordless
  • Supplier No.:
    215
Specifications
About this item
These affordable microscopes include a number of high-quality features.
  • Wide-Field 10x Eyepiece with Pointer
  • Bulb Life: 10,000 Hours
  • Four-Hole Reversed Objective Turret
Features include a wide-field 10x eyepiece with pointer and a four-hole reversed objective turret.