To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Environmental Chemistry: Acid Rain, Weathering, and Erosion
- Cat. no.:470136-734
- Supplier No.:IS9703
Demonstrates the Effects of Acid Rain
Acid rain is a term used generically to describe any type of acidic moisture, whether rain, snow, or fog. It can have devastating effects on aquatic ecosystems, as well as terrestrial areas, buildings, statues, and monuments. In this activity, students expose many rock materials and metals, both naturally occurring and common in construction, to an acidic environment and examine the reaction of these materials in contrast to the same materials exposed to “normal” rain (tap water). Kit contains materials for 15 groups, Teacher’s Manual, Student Study Guide, and copymasters.