Omni Tissue Master 125 Handheld Homogenizers, Omni International, Inc.
Fournisseur: Omni International
Certificats
À propos de cet article
The economically priced Tissue Master 125 is well suited for customers who homogenize with little variation in sample size
The Tissue Master is an ideal choice for a laboratory with many applications and a limited budget. Recommended for soft tissue applications and emulsions only.
It comes complete with a 5mm, 7mm, and 10mm stainless steel generator probe that can handle a variety of different types of samples.
Certifications: CE Certified.
Ordering information: Includes 125 watt motor, stainless steel generator probe, 1-year motor warranty & user manual.
Spécifications
- Catalog No:
- CA11010-492
- CA11010-488
- Speed range:
- 5,000-35,000 rpm
- 5,000-35,000 rpm
- Weight:
- 535 g (18.8 oz.)
- 535 g (18.8 oz.)
- Power rating:
- 125 W
- 125 W