Flanging Tool Kit, Hamilton
Flanging Tool Kit, Hamilton
Hamilton Flanging Tool Kit, 120 V
Catalog # 65514-003
Supplier:   Hamilton
Supplier #:  88821
Flanging Tool Kit, Hamilton
Catalog # 65514-003
Supplier:  Hamilton
Flanging Tool Kit, Hamilton
Hamilton Flanging Tool Kit, 120 V
Price
$1,119.42
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Hamilton Company offers the most comprehensive selection of standard and custom syringes and accessories in the industry.
The flanging tool consists of a soldering iron and custom heating tips. The custom tips are ideally designed to apply heat to a squarely cut piece of tubing. As the tubing is heated it is formed into a flange that is ideally suited for sealing into Hamilton valves. The kit comes with 3 heating tips for 12, 18, and 22 gauge tubing as well as a tubing holder. These components are also available individually. Each is manufactured to achieve the highest level of accuracy and precision possible.
 : If you are looking for a syringe, valve or accessory that is available but not listed here, please call Avantor to place a special order by phone.
Specifications
  • Désignation:
    Hamilton Flanging Tool Kit
  • Tension:
    120 V