JACKSON SAFETY® V20 PURITY™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Catalogue # CA89215-564
Fournisseur: Microflex
Spécifications
- Frame Color:Clear
- Cat. no.:CA89215-564
- Lens Coating:Anti-fog
- Lens Tint:Clear
Spécifications
À propos de cet article
Lightweight, 10.25-base curved lens provides wraparound protection for a wide field of vision.
- Universal nose bridge
- Polycarbonate lens provides 99.9% U.V. protection
Soft, non-slip padded temples ensure a comfortable fit.
Certifications: NIOSH approved.Meets ANSI Z87.1+ standards.