ContactUsLinkComponent
VWR® PCR-To-Gel Taq Master Mix, 2X
Catalog # 97068-148
Supplier:  VWR
CAS Number:  
undefined
VWR® PCR-To-Gel Taq Master Mix, 2X
Catalog # 97068-148
Supplier:  VWR
Supplier Number:  1B1409-100RXN
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Hot Start PCR-to-Gel Master Mix, 2X
  • Size:
    100 rxn
  • Cat. no.:
    97068-148
  • Volume:
    2× 1.25 ml
  • Supplier No.:
    1B1409-100RXN
Specifications
About this item
Hot Start PCR-To-Gel Taq Master Mix, 2X is a ready-to-use reaction cocktail that contains all required components, except primers and template DNA, for routine PCR amplification of DNA fragments up to 4 kb
The included Taq DNA polymerase is inactivated with proprietary monoclonal antibodies. Incubation at ≥ 94°C during PCR thermal cycling irreversibly denatures the antibody and releases full Taq DNA polymerase activity. Inclusion of tracking dyes in the master mix allows direct loading and analysis of PCR products by agarose gel electrophoresis following PCR amplification.