Staining Dish, 10 Slide Unit, Wheaton
10 Slide Staining Dish with Cover
Supplier: DWK Life Sciences (KIMBLE)
Fournisseur #: 900170
Jump to:
Product Details & Documents
Designed for staining 3x1" (75x25mm) microscope slides
Holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back-to-back. The dish and cover are manufactured from soda-lime glass. Approximate inside dimensions are 80Lx55Wx35D mm.
Spécifications
- Désignation:10 Slide Staining Dish with Cover