ContactUsLinkComponent
Phage Display Competent Cells, Biosearch Technologies
Phage Display Competent Cells, Biosearch Technologies
Supplier:  Lucigen
Certificates
undefined
Phage Display Competent Cells, Biosearch Technologies
Supplier:  Lucigen
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Catalog No:
  • 95057-476
  • 95057-470
  • 95040-450
  • 95040-452
  • CA95057-474
  • 95057-472
  • Genotypes:
  • ER2738: [F'proA+B+ lacIq Δ(lacZ)M15 zzf::Tn10 (tetr)] fhuA2 glnVΔ(lac-proAB) thi-1Δ(hsdS-mcrB)6
  • SS320 (MC1061F'): [F'proAB+lacIqlacZΔM15 Tn10 (tetr)] hsdR mcrB araD139 Δ(araABC-leu)7679ΔlacX74 galUgalK rpsL thi
  • TG1: [F' traD36 proAB lacIqZ ΔM15] supE thi-1 Δ(lac-proAB) Δ(mcrB-hsdSM)5(rK - mK -)
  • TG1: [F' traD36 proAB lacIqZ ΔM15] supE thi-1 Δ(lac-proAB) Δ(mcrB-hsdSM)5(rK - mK -)
  • ER2738: [F'proA+B+ lacIq Δ(lacZ)M15 zzf::Tn10 (tetr)] fhuA2 glnVΔ(lac-proAB) thi-1Δ(hsdS-mcrB)5
  • SS320 (MC1061F'): [F'proAB+lacIqlacZΔM15 Tn10 (tetr)] hsdR mcrB araD139 Δ(araABC-leu)7679ΔlacX74 galUgalK rpsL thi