Corning® Trypsin
Corning® Trypsin
 :   Corning
Corning® Trypsin
 :  Corning
Corning® Trypsin
Corning® Trypsin
Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging. Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances the efficacy of trypsin.
  • Storage: -25°C to -15°C
Trypsin has a pH specification range from 7.2 to 8. At this range, phenol red inclusion gives the solution a pink color. During shipping, the trypsin might change color from pink to yellow due to exposure to carbon dioxide released from the dry ice in the shipping container. In our experience, this discoloration does not affect the functionality of the trypsin. By adjusting the pH to 7.2 – 8 with NaOH the trypsin activity could be restored.
Product Family Options
  • Désignation
    Taille
  • Trypsin/EDTA Solution, 1x 0.05% Trypsin 0.53 mM EDTA in HBSS
    100 ml
    Pack of 1
    Pack of 1 - $105.00
    $105.00
    Pack of 1
       $105.00
    Minimum order quantity is 1
     : 25-051-CI
  • Trypsin/EDTA Solution, 1x 0.05% Trypsin 0.53 mM EDTA in HBSS
    100 ml
    Pack of 6 (100ml)
    Pack of 6 (100ml) - $96.20
    $96.20
    Pack of 6 (100ml)
       $96.20
     : 25-052-CI
  • Trypsin/EDTA Solution,1x 0.25% Trypsin 2.21 mM EDTA in HBSS
    100 ml
    Pack of 6 (100ml)
    Pack of 6 (100ml) - $99.85
    $99.85
    Pack of 6 (100ml)
       $99.85
     : 25-053-CI
  • Trypsin/EDTA Solution, 1x 0.05% Trypsin with 0.53 mM EDTA
    500 ml
    Pack of 6 (500ml)
    Pack of 6 (500ml) - $403.00
    $403.00
    Pack of 6 (500ml)
       $403.00
    Minimum order quantity is 1
     : 25-052-CV