Corning® S.O.C Medium, Corning | Avantor
Corning® S.O.C Medium, Corning
S.O.C Medium
Catalog # CA45001-098
Supplier:   Corning
Fournisseur #:  46-003-CR
Corning® S.O.C Medium, Corning
Catalog # CA45001-098
Supplier:  Corning
CAS Number:  
Corning® S.O.C Medium, Corning
Corning® S.O.C Medium, Corning
S.O.C Medium
CAS Number:  
Price
$146.00
Log in pour la disponibilité
Pack of 10 (10ml)
Product Details & Documents
S.O.C
The use of SOC medium improves transformation efficiency.
medium is used as a recovery medium during the final stage of transformation of E. coli.
Spécifications
Spécifications
  • Désignation:
    S.O.C Medium