Corning® Lymphocyte Separation Medium, Corning
Corning® Lymphocyte Separation Medium, Corning
Supplier:   Corning
Corning® Lymphocyte Separation Medium, Corning
Supplier:  Corning
Certificates
Corning® Lymphocyte Separation Medium, Corning
Corning® Lymphocyte Separation Medium, Corning
2 Options Available Below
Product Details & Documents
Lymphocyte Separation Medium (LSM) was originally designed for the in vitro isolation of lymphocytes from diluted whole blood
It is a sterile, iso-osmotic polysucrose and diatrizoate solution with low viscosity and a density of 1.077-1.080 g/mL at 20ºC.
Specifications for Product Family
Specifications
  • Catalog No:
  • CA45000-726
  • CA45000-728
  • Shipping conditions:
  • Ambient
  • Ambient
  • Storage conditions:
  • Ambient
  • Ambient
Specifications
Product Family Options
Product Information
  • Désignation
    Volume
    Availability
    Price
  • Lymphocyte Separation Medium
    100 ml
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $59.20
    $59.20
    Each (100ml)
       $59.20
    Supplier Number: 25-072-CI
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-726
    Shipping conditions
    Ambient
    Storage conditions
    Ambient
  • Lymphocyte Separation Medium
    500 ml
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $217.00
    $217.00
    Each (500ml)
       $217.00
    Supplier Number: 25-072-CV
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-728
    Shipping conditions
    Ambient
    Storage conditions
    Ambient