Corning® Cellstripper® Liquid, Corning | Avantor
Corning® Cellstripper® Liquid, Corning
Corning® Cellstripper® Liquid, 100 ml
Catalog # CA45000-668
Supplier:   Corning
Fournisseur #:  25-056-CI
Corning® Cellstripper® Liquid, Corning
Catalog # CA45000-668
Supplier:  Corning
CAS Number:  
Corning® Cellstripper® Liquid, Corning
Corning® Cellstripper® Liquid, Corning
Corning® Cellstripper® Liquid, 100 ml
CAS Number:  
Price
$191.35
Log in pour la disponibilité
Pack of 6 (100ml)
La quantité minimale de commande est de 1
Product Details & Documents
Cellstripper® is a non-enzymatic cell dissociation solution formulated with a proprietary mixture of chelators that gently dislodge adherent cells in tissue culture. It is an alternative to trypsin when used with either serum-free or serum-supplemented media. Because Cellstripper® is a gentler reagent than trypsin, cells can be exposed to Cellstripper for longer periods of time. Cellstripper® achieves superior results in comparison to enzymatic dissociation products.
Spécifications
Spécifications
  • Désignation:
    Corning® Cellstripper® Liquid
  • Volume:
    100 ml
  • Cat. No.:
    CA45000-668
  • Storage temperature:
    Ambient
  • Shipping temperature:
    Ambient